Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRP54 Recombinant Rabbit Monoclonal Antibody (1O3J1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542773
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, Jurkat. Immunogen sequence: PGRIQRVARG SGVSTRDVQE LLTQYTKFAQ MVKKMGGIKG LFKGGDMSKN VSQSQMAKLN QQMAKMMDPR VLHHMGGMAG LQSMMRQFQQ GAAGNMKGMM GFNNM.
Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).
Specifications
SRP54 | |
Recombinant Monoclonal | |
0.4 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61011, Q6AYB5 | |
SRP54, Srp54a | |
A synthetic peptide corresponding to a sequence within amino acids 400-504 of human SRP54 (P61011). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
1O3J1 | |
Unconjugated | |
SRP54 | |
54K subunit of signal recognition particle; 54kDa; signal recognition particle 54; signal recognition particle 54 kDa protein; signal recognition particle 54A; signal recognition particle 54kD; signal recognition particle 54kDa; SRP54; Srp54a | |
Rabbit | |
Affinity chromatography | |
RUO | |
116650, 24067, 6729 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction