Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRp55 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204142DL594
Description
SRp55 Polyclonal antibody specifically detects SRp55 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SRp55 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
B52, FLJ08061, pre-mRNA splicing factor SRP55, Pre-mRNA-splicing factor SRP55, serine/arginine-rich splicing factor 6, SFRS6, Splicing factor, arginine/serine-rich 6MGC5045, splicing factor, arginine/serine-rich, 55 kDa, SR splicing factor 6, SRP55arginine/serine-rich splicing factor 6 | |
The immunogen recognized by this antibody maps to this antibody using the numbering given in entry NP_006266.2 (GeneID 6431). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6431 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction