Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRP68 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100080 Supplier Thermo Scientific Supplier No. PA5100080
Add to Cart
Add to Cart

SRP68 Polyclonal Antibody

SRP68 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP68
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SRP68
Gene Accession No. Q8BMA6, Q9UHB9
Gene Alias 2610024I03Rik; 68kDa signal recognition particle; 68kDA subunit of signal recognition particle; likely ortholog of H. sapiens signal recognition particle 68kDa (SRP68); signal recognition particle 68; Signal recognition particle 68 kDa protein; signal recognition particle 68kD; signal recognition particle 68kDa; signal recognition particle subunit SRP68; Srp68; zgc:92573
Gene Symbols SRP68
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRP68(Accession Q9UHB9), corresponding to amino acid residues P553-L603.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217337, 363707, 6730
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.