Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRP68 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554752 Supplier Invitrogen™ Supplier No. PA554752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAKTKLEAQA YTAYLSGMLR FEHQEWKAAI EAFNKCKTIY EKLASAFTEE QAVLYNQRVE EISPNIRYCA YNIGDQSAIN ELMQMRLRSG GTEGLLAEKL EALITQTRAK QAATMSEVEW RGRTVPVKID KVRIFLLGLA DNEAAIV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The signal recognition particle (SRP) is a ribonucleoprotein complex that transports secreted and membrane proteins to the endoplasmic reticulum for processing. The complex includes a 7S RNA and six protein subunits. SRP68 is the 68kDa component of the SRP.The signal recognition particle (SRP) is a ribonucleoprotein complex that transports secreted and membrane proteins to the endoplasmic reticulum for processing. The complex includes a 7S RNA and six protein subunits. This gene encodes the 68kDa component of the SRP. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined. Three related pseudogenes are located within the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP68
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRP68
Gene Accession No. Q9UHB9
Gene Alias 2610024I03Rik; 68kDa signal recognition particle; 68kDA subunit of signal recognition particle; likely ortholog of H. sapiens signal recognition particle 68kDa (SRP68); signal recognition particle 68; Signal recognition particle 68 kDa protein; signal recognition particle 68kD; signal recognition particle 68kDa; signal recognition particle subunit SRP68; Srp68; zgc:92573
Gene Symbols SRP68
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRP68. Recombinant protein control fragment (Product #RP-93391).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6730
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.