Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRP72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143946
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143946 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143946 Supplier Invitrogen™ Supplier No. PA5143946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human K562 whole cell, human HL-60 whole cell, rat pancreas tissue, mouse pamcreas tissue. IHC: human breast cancer tissue, human renal cancer tissue, human thyroid cancer tissue, human liver cancer tissue, mouse brain tissue, rat brain tissue. ICC/IF: U20S cell. Flow: U937 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. Binds the 7S RNA only in presence of SRP68. This ribonucleoprotein complex might interact directly with the docking protein in the ER membrane and possibly participate in the elongation arrest function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP72
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene SRP72
Gene Accession No. O76094
Gene Alias 5730576P14Rik; 72kDa; AI132477; BC019196; BMFF; BMFS1; C77589; epididymis luminal protein 103; HEL103; signal recognition particle 72; signal recognition particle 72 kDa; signal recognition particle 72 kDa protein; signal recognition particle 72kD; signal recognition particle 72kDa; signal recognition particle subunit SRP72; SRP72
Gene Symbols SRP72
Host Species Rabbit
Immunogen E.coli-derived human SRP72 recombinant protein (Position: I40-Y460).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498351, 66661, 6731
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.