Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRP72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112246 Supplier Invitrogen™ Supplier No. PA5112246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRP72 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. Binds the 7S RNA only in presence of SRP68. This ribonucleoprotein complex might interact directly with the docking protein in the ER membrane and possibly participate in the elongation arrest function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP72
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene SRP72
Gene Accession No. O76094
Gene Alias 5730576P14Rik; 72kDa; AI132477; BC019196; BMFF; BMFS1; C77589; epididymis luminal protein 103; HEL103; signal recognition particle 72; signal recognition particle 72 kDa; signal recognition particle 72 kDa protein; signal recognition particle 72kD; signal recognition particle 72kDa; signal recognition particle subunit SRP72; SRP72
Gene Symbols SRP72
Host Species Rabbit
Immunogen E. coli-derived Human SRP72 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.