Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRP9 Mouse anti-Human, Mouse, Rat, Clone: 1H7G10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | SRP9 |
---|---|
Clone | 1H7G10 |
Concentration | 0.93 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6505
![]() ![]() |
Proteintech Group Inc
660681IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.Specifications
SRP9 | |
0.93 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALURBP, SRP9 | |
SRP9 | |
IgG2a | |
Protein A | |
Antibody |
1H7G10 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P49458, P49962 | |
27058, 6726, 690345 | |
SRP9 Fusion Protein Ag17153 | |
Primary | |
-20°C | |
SRP9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title