Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRPK1 Monoclonal Antibody (03)
GREENER_CHOICE

Catalog No. PIMA530905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530905 Supplier Invitrogen™ Supplier No. MA530905
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SRPK1.

SRPK1 is a serine/threonine kinase that plays a role in the regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. SRPK1 localizes to the nucleus and the cytoplasm. It is thought to play a role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. A second alternatively spliced transcript variant for this gene has been described, but its full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPK1
Applications ELISA
Classification Monoclonal
Clone 03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SRPK1
Gene Accession No. Q96SB4
Gene Alias AU017960; OTTHUMP00000016277; serine/arginine-rich protein specific kinase 1; serine/arginine-rich protein-specific kinase 1; serine/arginine-rich splicing factor kinase 1; serine/threonine-protein kinase SRPK1; SFRS protein kinase 1; SFRSK1; SR protein-specific kinase 1; SRPK1; SR-protein-specific kinase 1; SRSF protein kinase 1
Gene Symbols SRPK1
Host Species Mouse
Immunogen Recombinant Human SRPK1 Protein (Glu2-Ser655).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6732
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.