Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRPK2 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297313APC
Description
SRPK2 Polyclonal antibody specifically detects SRPK2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SRPK2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EC 2.7.11.1, FLJ36101, H_RG152G17.1a, H_RG152G17.1b, serine kinase SRPK2, Serine/arginine-rich protein-specific kinase 2, serine/arginine-rich splicing factor kinase 2, serine/threonine-protein kinase SRPK2, SFRS protein kinase 2SFRSK2, SR protein kinase 2, SR-protein-specific kinase 2, SRSF protein kinase 2, WUGSC:H_RG152G17.1a | |
Produced in rabbits immunized with E. coli-derived Human SRPK2 fragment. | |
0.1 mL | |
Protein Kinase | |
6733 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction