Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRPK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528654 Supplier Invitrogen™ Supplier No. PA528654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Molt-4, mouse brain. Predicted reactivity: Mouse (93%), Rat (91%), Rhesus Monkey (99%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SRPK2, along with SRPK1, is most likely the cellular protein kinase mediating HBV core protein phosphorylation during viral infection. It therefore represents an important host cell target for therapeutic intervention in HBV infection. Promotes neuronal apoptosis by up-regulating cyclin-D1 (CCND1) expression. This is done by the phosphorylation of SRSF2, leading to the suppression of p53/TP53 phosphorylation thereby relieving the repressive effect of p53/TP53 on cyclin-D1 (CCND1) expression. Phosphorylates ACIN1, and redistributes it from the nuclear speckles to the nucleoplasm, resulting in cyclin A1 but not cyclin A2 up-regulation. Plays an essential role in spliceosomal B complex formation via the phosphorylation of DDX23/PRP28. Plays a negative role in the regulation of HBV replication through a mechanism not involving the phosphorylation of the core protein but by reducing the packaging efficiency of the pregenomic RNA (pgRNA) without affecting the formation of the viral core particles .
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPK2
Applications Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SRPK2
Gene Accession No. O54781, P78362
Gene Alias AW226533; AW492537; AW547358; H_RG152G17.1a; H_RG152G17.1b; mSRPK2; serine kinase SRPK2; serine/arginine-rich protein specific kinase 2; serine/arginine-rich protein-specific kinase 2; serine/arginine-rich splicing factor kinase 2; serine/threonine-protein kinase SRPK2; SFRS protein kinase 2; SFRSK2; SR protein kinase 2; SRPK2; SR-protein-specific kinase 2; SRSF protein kinase 2; SRSF protein kinase 2 C-terminal; SRSF protein kinase 2 N-terminal; Wbp6; WUGSC:H_RG152G17.1a; WW domain binding protein 6
Gene Symbols SRPK2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SRPK2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20817, 6733
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.