Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRPR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541770
Description
Peptide sequence: DSEKAKKPVR SMIETRGEKP KEKAKNSKKK GAKKEGSDGP LATSKPVPAE Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 93%.
The gene encodes a subunit of the endoplasmic reticulum signal recognition particle receptor that, in conjunction with the signal recognition particle, is involved in the targeting and translocation of signal sequence tagged secretory and membrane proteins across the endoplasmic reticulum. Alternative splicing results in multiple transcript variants.
Specifications
| SRPR | |
| Polyclonal | |
| Unconjugated | |
| Srpra | |
| 1300011P19Rik; D11Mgi27; Docking protein alpha; DP; DP-alpha; MGC17355; MGC3650; MGC9571; signal recognition particle receptor (docking protein); signal recognition particle receptor ('docking protein'); signal recognition particle receptor subunit alpha; Sralpha; SR-alpha; SRP receptor alpha subunit; SRP receptor subunit alpha; SRP-alpha; Srpr; SRPRA; Unknown (protein for MGC:128907) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6734 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P08240 | |
| Srpra | |
| Synthetic peptide directed towards the N-terminal of human SRPR (aa 141-190). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction