Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRPR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117035 Supplier Invitrogen™ Supplier No. PA5117035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, HT-1080 Immunogen sequence: MLDFFTIFSK GGLVLWCFQG VSDSCTGPVN ALIRSVLLQE RGGNNSFTHE ALTLKYKLDN QFELVFVVGF QKILTLTYVD KLIDDVHRLF RDKYRTEIQQ QSALSLLNGT FDFQNDFLRL LREAEESSKI RAPTTMKKFE DSEKAKKPVR SMIETRGEKP KEKAKNSKKK GAKKEGSDGP LATSKPVPAE KSGLPVGPEN GVELSKEELI RRKREEFIQK.

The gene encodes a subunit of the endoplasmic reticulum signal recognition particle receptor that, in conjunction with the signal recognition particle, is involved in the targeting and translocation of signal sequence tagged secretory and membrane proteins across the endoplasmic reticulum. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPR
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Srpra
Gene Accession No. P08240
Gene Alias 1300011P19Rik; D11Mgi27; Docking protein alpha; DP; DP-alpha; MGC17355; MGC3650; MGC9571; signal recognition particle receptor (docking protein); signal recognition particle receptor ('docking protein'); signal recognition particle receptor subunit alpha; Sralpha; SR-alpha; SRP receptor alpha subunit; SRP receptor subunit alpha; SRP-alpha; Srpr; SRPRA; Unknown (protein for MGC:128907)
Gene Symbols Srpra
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human SRPR (NP_0031302).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6734
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.