Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRPRB Antibody, Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NBP266584
Description
SRPRB Polyclonal specifically detects SRPRB in Canine, Chicken samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SRPRB | |
Polyclonal | |
Western Blot 15 ug/ml, Immunohistochemistry 1:10 - 1:500 | |
APMCF1, Protein APMCF1, signal recognition particle receptor subunit beta, signal recognition particle receptor, B subunit, signal recognition particle receptor, beta subunit, SR-beta | |
Sheep | |
Ammonium sulfate precipitation | |
Primary | |
Antibody will not detect SRPRB protein bound to SRPRA | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
PBS with 0.08% Sodium Azide | |
SRPRB | |
Synthetic peptide corresponding to amino acids 246-265 derived from the carboxyl terminal of the canine SRPRB subunit. | |
250 μg | |
58477 | |
Canine, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction