Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRPRB Antibody (OTI2D4), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274351FR
Description
SRPRB Monoclonal specifically detects SRPRB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SRPRB | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
SRPRB | |
Full length recombinant protein of human SRPRB (NP_067026) produced in HEK293T cells. | |
0.1 mL | |
58477.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2D4 | |
Western Blot, Immunohistochemistry | |
APMCF1, Protein APMCF1, signal recognition particle receptor subunit beta, signal recognition particle receptor, B subunit, signal recognition particle receptor, beta subunit, SR-beta | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction