Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRPRB Monoclonal Antibody (OTI1D9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504712S


Description
SRPRB Monoclonal antibody specifically detects SRPRB in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
SRPRB | |
Monoclonal | |
0.88 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96CM8 | |
Mouse | |
Affinity chromatography | |
RUO | |
80221 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1D9 | |
Unconjugated | |
Acsf2 | |
AA409543; Ab2-417; APMCF1; Ba1-667; Cc1-8; hypothetical protein; Protein APMCF1; PSEC0230; Signal recognition particle receptor subunit beta; signal recognition particle receptor subunit beta pseudogene; signal recognition particle receptor, B subunit; signal recognition particle receptor, beta subunit; SR-beta; SRP receptor beta subunit; SRP receptor subunit beta; SRPRB; wu:fk57h10; zgc:111820; zgc:92746; zgc:92746 protein | |
Full length human recombinant protein of human ACSF2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction