Learn More
Invitrogen™ SRPRB Polyclonal Antibody

Description
Immunogen sequence: IRSRRSSQRA VLLVGLCDSG KTLLFVRLLT GLYRDTQTSI TDSCAVYRVN NNRGNSLTLI DLPGHESLRL QFLERFKSSA RAIVFVVDSA AFQREVKDVA EFLYQVLIDS MGLKNTPSFL IACNKQDIAM AKSAKLIQQQ LEKELNTLRV TRSAAPSTLD SSSTAPAQLG KKGKEFEFSQ LPLKVEFLEC SAKGGRGDVG SADIQDLEKW LAKIA; Positive Samples: MCF7, HT-29, LO2, , B cells, Mouse liver, Mouse pancreas, Mouse small intestine, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Single-pass membrane protein.
Specifications
Specifications
| Antigen | SRPRB |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.88 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | SRPRB |
| Gene Accession No. | P47758, Q4FZX7, Q9Y5M8 |
| Gene Alias | AA409543; Ab2-417; APMCF1; Ba1-667; Cc1-8; hypothetical protein; Protein APMCF1; PSEC0230; Signal recognition particle receptor subunit beta; signal recognition particle receptor subunit beta pseudogene; signal recognition particle receptor, B subunit; signal recognition particle receptor, beta subunit; SR-beta; SRP receptor beta subunit; SRP receptor subunit beta; SRPRB; wu:fk57h10; zgc:111820; zgc:92746; zgc:92746 protein |
| Gene Symbols | SRPRB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.