Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRPRB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587107
Description
Purity is > 95% by SDS-PAGE.
SRPRB has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane. The protein encoded by this gene has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane. The protein encoded by this gene has similarity to mouse protein which is a subunit of the signal recognition particle receptor (SR). This subunit is a transmembrane GTPase belonging to the GTPase superfamily. It anchors alpha subunit, a peripheral membrane GTPase, to the ER membrane. SR is required for the cotranslational targeting of both secretory and membrane proteins to the ER membrane.
Specifications
SRPRB | |
Polyclonal | |
Unconjugated | |
SRPRB | |
AA409543; Ab2-417; APMCF1; Ba1-667; Cc1-8; hypothetical protein; Protein APMCF1; PSEC0230; Signal recognition particle receptor subunit beta; signal recognition particle receptor subunit beta pseudogene; signal recognition particle receptor, B subunit; signal recognition particle receptor, beta subunit; SR-beta; SRP receptor beta subunit; SRP receptor subunit beta; SRPRB; wu:fk57h10; zgc:111820; zgc:92746; zgc:92746 protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20818, 300965, 58477 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P47758, Q4FZX7, Q9Y5M8 | |
SRPRB | |
Recombinant fusion protein of human SRPRB. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction