Learn More
Invitrogen™ SRPX Polyclonal Antibody

Description
A suggested positive control is SK-N-SH cell lysate. PA5-20843 can be used with blocking peptide PEP-0957.
Specifications
Specifications
| Antigen | SRPX |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SRPX |
| Gene Accession No. | P78539, Q63769, Q9R0M3 |
| Gene Alias | down-regulated by v-src; Drs; DRS protein; drs-1; drs-2; epididymis secretory sperm binding protein Li 83p; ETX1; HEL-S-83p; mDRS-1; mDRS-2; SRPX; SRPX1; sushi repeat containing protein, X-linked; sushi repeat-containing protein SRPX; sushi-repeat containing protein, X-linked; sushi-repeat-containing protein; sushi-repeat-containing protein SRPX; sushi-repeat-containing protein X chromosome; sushi-repeat-containing protein, X chromosome; sushi-repeat-containing protein, X-chromosome; sushi-repeat-containing protein, X-linked |
| Gene Symbols | SRPX |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.