Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRPX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116545 Supplier Invitrogen™ Supplier No. PA5116545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse heart, Mouse liver Immunogen sequence: EDDEVGYSHP RYKDTPWCSP IKVKYGDVYC RAPQGGYYKT ALGTRCDIRC QKGYELHGSS LLICQSNKRW SDKVICKQKR CPTLAMPANG GFKCVDGAYF NSRCEYYCSP GYTLKGERTV TCMDNKAWSG RPASCVDMEP PRIKCPSVKE RIAEPNKLTV RVSWETPEGR DTADGILTDV ILKGLPPGSN FPEGDHKIQY TVYDRAENKG TCKFRVKVRV KRCGKLNAPE NGYMKCSSDG DNYGATCEFS CIGGYELQGS PARVCQSNLA WSGTEPTCAA M.

SRPX1 was initially identified as a candidate gene for X-linked retinitis pigmentosa and is overexpressed in the trabecular network of the eye in glaucoma conditions. Its expression is significantly downregulated in a number cancer cell lines and malignant tumor tissues and its ectopic expression induces apoptosis through the endoplasmic reticulum and the activation of caspase-12, -9, and -3, indicating that it can function as a tumor suppressor. SRPX1 is also involved with the regulation of autophagy under low serum conditions and can associate with Rab24, a member of the Rab GTPase protein family that is involved in autophagy. This association of SRPX1 with Rab24 is enhanced during autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPX
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SRPX
Gene Accession No. Q9R0M3
Gene Alias down-regulated by v-src; Drs; DRS protein; drs-1; drs-2; epididymis secretory sperm binding protein Li 83p; ETX1; HEL-S-83p; mDRS-1; mDRS-2; SRPX; SRPX1; sushi repeat containing protein, X-linked; sushi repeat-containing protein SRPX; sushi-repeat containing protein, X-linked; sushi-repeat-containing protein; sushi-repeat-containing protein SRPX; sushi-repeat-containing protein X chromosome; sushi-repeat-containing protein, X chromosome; sushi-repeat-containing protein, X-chromosome; sushi-repeat-containing protein, X-linked
Gene Symbols SRPX
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-320 of human SRPX (NP_0062981).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51795
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.