Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRPX2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520844

Catalog No. PIPA520844


Only null left
Add to Cart

Description

Description

A suggested positive control is human lung tissue lysate. PA5-20844 can be used with blocking peptide PEP-0958.

Sushi-repeat-containing protein X-linked 2 (SRPX2) is a neural gene functioning in the speech and language center of the human brain; mutations in this gene lead to epilepsy, speech dyspraxia, mental retardation and cognitive disorders. Recently, SRPX2 was found to be a novel mediator of angiogenesis and can act as a ligand for the urokinase-type plasminogen activator, a protein that can facilitate invasive migration of sprouting endothelial cells. SRPX2 is also overexpressed in gastric cancer, leading to increased phosphorylation levels of focal adhesion kinase and enhanced cellular migration and adhesion, suggesting that SRPX2 may be a potential target in the treatment of metastatic cancers.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SRPX2
Polyclonal
Unconjugated
SRPX2
1110039C07Rik; BPP; CBPS; EMBL:AAI68975.1, ECO:0000312; PMGX; PubMed:24179158}; RESDX; RGD:1562444}; RGD1562444; RP11-524D16__A.1; SRP; SRPUL; SRPX2; srpx2 {ECO:0000312; sushi repeat containing protein X-linked 2; sushi repeat containing protein, X-linked 2; sushi repeat-containing protein SRPX2; sushi repeat-containing protein SRPX2 {ECO:0000303; sushi-repeat containing protein; sushi-repeat containing protein, X-linked 2; sushi-repeat protein; sushi-repeat protein upregulated in leukemia; sushi-repeat protein up-regulated in leukemia; sushi-repeat-containing protein, X-linked 2
Rabbit
Antigen affinity chromatography
RUO
27286, 317181, 68792
4°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
B5DF94, O60687, Q8R054
SRPX2
A 16 amino acid peptide from near the amino terminus of human SRPX2.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.