Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRPX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209251
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9251 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9251 Supplier Fabgennix Inc Supplier No. SRPX2201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRPX2 Polyclonal antibody specifically detects SRPX2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen SRPX2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene SRPX2
Gene Accession No. B5DF94, O60687, Q8R054
Gene Alias 1110039C07Rik; BPP; CBPS; EMBL:AAI68975.1, ECO:0000312; PMGX; PubMed:24179158}; RESDX; RGD:1562444}; RGD1562444; RP11-524D16__A.1; SRP; SRPUL; SRPX2; srpx2 {ECO:0000312; sushi repeat containing protein X-linked 2; sushi repeat containing protein, X-linked 2; sushi repeat-containing protein SRPX2; sushi repeat-containing protein SRPX2 {ECO:0000303; sushi-repeat containing protein; sushi-repeat containing protein, X-linked 2; sushi-repeat protein; sushi-repeat protein upregulated in leukemia; sushi-repeat protein up-regulated in leukemia; sushi-repeat-containing protein, X-linked 2
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 430-470 on human SRPX2 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27286, 317181, 68792
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.