Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104123
Description
Antibody detects endogenous levels of total SRR.
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
Specifications
SRR | |
Polyclonal | |
Unconjugated | |
Srr | |
D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs | |
Rabbit | |
Affinity chromatography | |
RUO | |
27364, 303306, 63826 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q76EQ0, Q9GZT4, Q9QZX7 | |
Srr | |
A synthesized peptide derived from human SRR(Accession Q9GZT4), corresponding to amino acid residues D112-G162. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction