Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112725
Description
SRR Polyclonal Antibody for Western Blot, IHC (P), ELISA
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
Specifications
| SRR | |
| Polyclonal | |
| Unconjugated | |
| Srr | |
| D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs | |
| Rabbit | |
| Protein G | |
| RUO | |
| 27364, 63826 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9GZT4, Q9QZX7 | |
| Srr | |
| Recombinant Human Serine racemase protein (1-340AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction