Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRRM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596499 Supplier Invitrogen™ Supplier No. PA596499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-29, OVCAR-3, LO2, HeLa, Jurkat, Mouse liver, Mouse kidney; Cellular Location: Nucleus matrix, Nucleus speckle.

Part of pre- and post-splicing multiprotein mRNP complexes. Involved in numerous pre-mRNA processing events. Promotes constitutive and exonic splicing enhancer (ESE)-dependent splicing activation by bridging together sequence-specific (SR family proteins, SFRS4, SFRS5 and TRA2B/SFRS10) and basal snRNP (SNRP70 and SNRPA1) factors of the spliceosome. Stimulates mRNA 3'-end cleavage independently of the formation of an exon junction complex. Binds both pre-mRNA and spliced mRNA 20-25 nt upstream of exon-exon junctions. Binds RNA and DNA with low sequence specificity and has similar preference for either double- or single-stranded nucleic acid substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRRM1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SRRM1
Gene Accession No. Q52KI8, Q8IYB3
Gene Alias 160-KD; AA407769; LOW QUALITY PROTEIN: serine/arginine repetitive matrix protein 1; plenty-of-prolines 101; plenty-of-prolines-101; Pop101; Ser/Arg-related nuclear matrix protein; serine and arginine repetitive matrix 1; serine/arginine repetitive matrix 1; serine/arginine repetitive matrix protein 1; Srm160; SR-related nuclear matrix protein of 160 kDa; SRRM1; wu:fb34f04; wu:fb38b06; zgc:63802
Gene Symbols SRRM1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10250, 51796
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.