Learn More
Invitrogen™ SRRM2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SRRM2.
Specifications
Specifications
| Antigen | SRRM2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SRRM2 |
| Gene Accession No. | Q8BTI8, Q9UQ35 |
| Gene Alias | 300 kDa nuclear matrix antigen; 300-KD; 5033413A03Rik; AA410130; Cwc21; CWF21; HSPC075; Kiaa0324; mKIAA0324; RNA binding protein; ser/Arg-related nuclear matrix protein of 300 kDa; serine/arginine repetitive matrix 2; serine/arginine repetitive matrix protein 2; Serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa; splicing coactivator subunit SRm300; SRL300; SRm300; SR-related nuclear matrix protein of 300 kDa; SRRM2; taxREB803; Tax-responsive enhancer element-binding protein 803; testicular secretory protein Li 53 |
| Gene Symbols | SRRM2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.