Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRRM2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988247
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8247 Supplier Bioss Supplier No. BS8067R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRRM2 is involved in pre-mRNA splicing. It may function at or prior to the first catalytic step of splicing at the catalytic center of the spliceosome and may do so by stabilizing the catalytic center or the position of the RNA substrate. It binds to RNA.

Specifications

Antigen SRRM2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SRRM2
Gene Accession No. Q9UQ35
Gene Alias 300 kDa nuclear matrix antigen; 300-KD; 5033413A03Rik; AA410130; BOS_22611; Cwc21; CWF21; HSPC075; Kiaa0324; LOC100340621; mKIAA0324; RNA binding protein; ser/Arg-related nuclear matrix protein of 300 kDa; serine/arginine repetitive matrix 2; Serine/arginine repetitive matrix protein 2; Serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa; Splicing coactivator subunit SRm300; splicing coactivator subunit SRm300-like; SRL300; SRM300; SR-related nuclear matrix protein of 300 kDa; Srrm2; TaxREB803; Tax-responsive enhancer element-binding protein 803; testicular secretory protein Li 53
Gene Symbols SRRM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SRRM2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23524
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.