Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRRM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112589
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112589 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112589 Supplier Invitrogen™ Supplier No. PA5112589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRRM4 Polyclonal Antibody for ELISA, IHC (P), Western Blot

SRRM4 promotes alternative splicing and inclusion of neural-specific exons in target mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRRM4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SRRM4
Gene Accession No. A7MD48, Q8BKA3
Gene Alias 1500001A10Rik; B230202K19Rik; bv; KIAA1853; Medulloblastoma antigen MU-MB-2.76; MGC176729; mKIAA1853; MU-MB-2.76; neural-specific serine/arginine repetitive splicing factor of 100 kDa; Neural-specific SR-related protein of 100 kDa; nSR100; serine/arginine repetitive matrix 4; serine/arginine repetitive matrix protein 4; SRRM2C; SRRM4
Gene Symbols SRRM4
Host Species Rabbit
Immunogen Recombinant Human Serine/arginine repetitive matrix protein 4 protein (400-479AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498189, 68955, 84530
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.