Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRRM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545083
Description
Peptide sequence: KTASPLTTSR GRSQEYDSGN DTSSPPSTQT SSARSRGQEK GSPSGGLSKS Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.
SRRM4 promotes alternative splicing and inclusion of neural-specific exons in target mRNAs.
Specifications
| SRRM4 | |
| Polyclonal | |
| Unconjugated | |
| SRRM4 | |
| 1500001A10Rik; B230202K19Rik; bv; KIAA1853; Medulloblastoma antigen MU-MB-2.76; MGC176729; mKIAA1853; MU-MB-2.76; neural-specific serine/arginine repetitive splicing factor of 100 kDa; Neural-specific SR-related protein of 100 kDa; nSR100; serine/arginine repetitive matrix 4; serine/arginine repetitive matrix protein 4; SRRM2C; SRRM4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84530 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Surface Plasmon Resonance | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A7MD48 | |
| SRRM4 | |
| Synthetic peptide directed towards the middle region of human SRRM4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction