Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRRM4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988249
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8249 Supplier Bioss Supplier No. BS8069R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRRM4 promotes alternative splicing and inclusion of neural-specific exons in target mRNAs.

Specifications

Antigen SRRM4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SRRM4
Gene Accession No. A7MD48
Gene Alias 1500001A10Rik; B230202K19Rik; bv; KIAA1853; Medulloblastoma antigen MU-MB-2.76; MGC176729; mKIAA1853; MU-MB-2.76; Neural-specific serine/arginine repetitive splicing factor of 100 kDa; Neural-specific SR-related protein of 100 kDa; nSR100; serine/arginine repetitive matrix 4; Serine/arginine repetitive matrix protein 4; SRRM2C; SRRM4
Gene Symbols SRRM4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SRRM4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84530
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.