Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRRM5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561501
Description
Immunogen sequence: SSKPSMSLAP SGSSMPTADP KPPASLKSTK SATPNRSLVP TKPATSRNSV MSPSSSKSTK STSTKRAPSN RPSSRSRVRS KARTPSRVST DTRTSKASKA SDVRCHQR Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 53%.

Specifications
SRRM5 | |
Polyclonal | |
Unconjugated | |
SRRM5 | |
serine/arginine repetitive matrix 5; serine/arginine repetitive matrix protein 5; SRRM5; zinc finger protein 576, isoform 2; ZNF576 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100170229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
B3KS81 | |
SRRM5 | |
Recombinant protein corresponding to Human SRRM5. Recombinant protein control fragment (Product #RP-100676). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction