Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRrp35 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15740520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15740520 20 μL
NBP157405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15740520 Supplier Novus Biologicals Supplier No. NBP15740520UL

Rabbit Polyclonal Antibody

SRrp35 Polyclonal specifically detects SRrp35 in Human samples. It is validated for Western Blot.

Specifications

Antigen SRrp35
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q8WXF0
Gene Alias 35 kDa SR repressor protein, FLJ14459, serine/arginine-rich splicing factor 12, serine-arginine repressor protein (35 kDa), SFRS13B, SFRS19FLJ41221, Splicing factor, arginine/serine-rich 13BFLJ33484, Splicing factor, arginine/serine-rich 19, SR splicing factor 12, SRRP35, SRrp35RP11-63L7.3
Gene Symbols SRSF12
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SRRP35 The peptide sequence was selected from the middle region of SRRP35. Peptide sequence RSRSKSLQKRSKSIGKSQSSSPQKQTSSGTKSRSHGRHSDSIARSPCKSP.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135295
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.