Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRrp35 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SRrp35 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SRrp35 Polyclonal specifically detects SRrp35 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SRrp35 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
135295 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: RSTSARQSRTPRRNFGSRGRSRSKSLQKRSKSIGKSQSSSPQKQTSSGTKSRSHGRHSDSIARS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
35 kDa SR repressor protein, FLJ14459, serine/arginine-rich splicing factor 12, serine-arginine repressor protein (35 kDa), SFRS13B, SFRS19FLJ41221, Splicing factor, arginine/serine-rich 13BFLJ33484, Splicing factor, arginine/serine-rich 19, SR splicing factor 12, SRRP35, SRrp35RP11-63L7.3 | |
SRSF12 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title