Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRrp35 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562044
Description
Immunogen sequence: RSTSARQSRT PRRNFGSRGR SRSKSLQKRS KSIGKSQSSS PQKQTSSGTK SRSHGRHSDS IARS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
SRSF12 gene ontology annotations related to this gene include regulation of alternative mRNA splicing, via spliceosome.
Specifications
SRrp35 | |
Polyclonal | |
Unconjugated | |
SRSF12 | |
35 kDa serine-arginine repressor; 35 kDa SR repressor protein; 35 kDa SR repressor protein-like protein; B830039L16; RGD1561162; RP11-63L7.3; serine and arginine rich splicing factor 12; serine/arginine-rich splicing factor 12; serine/arginine-rich splicing factor 13B; serine-arginine repressor protein; serine-arginine repressor protein (35 kDa); serine-arginine repressor protein 35; Sfrs13b; SFRS19; splicing factor, arginine/serine-rich 13B; Splicing factor, arginine/serine-rich 19; SR splicing factor 12; Srrp; SRRP35; Srsf12; Srsf13b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
135295 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WXF0 | |
SRSF12 | |
Recombinant protein corresponding to Human SRrp35. Recombinant protein control fragment (Product #RP-100421). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction