Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF1 Recombinant Rabbit Monoclonal Antibody (3J5Q9)
GREENER_CHOICE

Catalog No. PIMA535441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535441 Supplier Invitrogen™ Supplier No. MA535441
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: GGGGGGGAPR GRYGPPSRRS ENRVVVSGLP PSGSWQDLKD HMREAGDVCY ADVYRDGTGV VEFVRKEDMT YAVRKLDNTK FRSHEGETAY IRVKVDGPRS P.

UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 3J5Q9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3
Gene SRSF1
Gene Accession No. Q07955, Q6PDM2
Gene Alias 1110054N12Rik; 5730507C05Rik; 6330415C05Rik; AI482334; Alternative-splicing factor 1; ASF; ASF/SF2; ASF-1; AW491331; OK/SW-cl.3; pre-mRNA-splicing factor SF2, P33 subunit; pre-mRNA-splicing factor SRp30a; serine and arginine rich splicing factor 1; serine/arginine-rich splicing factor 1; SF2; SF2p33; SFRS1; splicing factor 2; splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich 1B; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1; SRp30a; Srsf1
Gene Symbols SRSF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SRSF1/SF2/ASF (NP_0088551).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110809, 6426, 689890
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.