Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF1 Recombinant Rabbit Monoclonal Antibody (JB40-76)
GREENER_CHOICE

Catalog No. PIMA534705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534705 Supplier Invitrogen™ Supplier No. MA534705
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Mouse heart tissue lysate, mouse liver tissue lysate, K562, A431, human lung cancer tissue, human tonsil tissue, human breast tissue, mouse colon tissue, rat epididymis tissue.

UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB40-76
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene SRSF1
Gene Accession No. Q07955
Gene Alias 1110054N12Rik; 5730507C05Rik; 6330415C05Rik; AI482334; Alternative-splicing factor 1; ASF; ASF/SF2; ASF-1; AW491331; OK/SW-cl.3; pre-mRNA-splicing factor SF2, P33 subunit; pre-mRNA-splicing factor SRp30a; serine and arginine rich splicing factor 1; serine/arginine-rich splicing factor 1; SF2; SF2p33; SFRS1; splicing factor 2; splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich 1B; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1; SRp30a; Srsf1
Gene Symbols SRSF1
Host Species Rabbit
Immunogen Synthetic peptide within Human SF2 aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6426
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.