Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114504
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114504 Supplier Invitrogen™ Supplier No. PA5114504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive controls that may be used include: K562 Cell Lysate.

SR proteins appear to be recruited from nuclear speckles, in which they are concentrated, to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SRSF3
Gene Accession No. P84103, P84104
Gene Alias AL024116; pre-mRNA splicing factor SRp20; pre-mRNA-splicing factor SRP20; Protein X16; serine and arginine rich splicing factor 3; serine/arginine-rich splicing factor 3; SFRS3; splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); splicing factor, arginine/serine-rich, 20-kD; Srp20; SRSF3; X16
Gene Symbols SRSF3
Host Species Rabbit
Immunogen SRSF3 antibody was raised against a 19 amino acid peptide near the center of human SRSF3. The immunogen is located within amino acids 80 - 130 of SRSF3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20383, 361814, 6428
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.