Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF3 Recombinant Rabbit Monoclonal Antibody (JE46-44)
GREENER_CHOICE

Catalog No. PIMA534846
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534846 Supplier Invitrogen™ Supplier No. MA534846
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Mouse testis tissue, rat brain tissue, A431, SH-SY-5Y, SiHa, 293, Hela, HepG2, A549, MG-63.

SR proteins appear to be recruited from nuclear 'speckles', in which they are concentrated, to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE46-44
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene SRSF3
Gene Accession No. P84103
Gene Alias AL024116; pre-mRNA splicing factor SRp20; pre-mRNA-splicing factor SRP20; Protein X16; serine and arginine rich splicing factor 3; serine/arginine-rich splicing factor 3; SFRS3; splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); splicing factor, arginine/serine-rich, 20-kD; Srp20; SRSF3; X16
Gene Symbols SRSF3
Host Species Rabbit
Immunogen Synthetic peptide within Human SFRS3 aa 34-83.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6428
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.