Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116691 Supplier Invitrogen™ Supplier No. PA5116691
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Mouse brain, Mouse kidney, Rat brain, Rat kidney Immunogen sequence: AEDAVRGLDG KVICGSRVRV ELSTGMPRRS RFDRPPARRP FDPNDRCYEC GEKGHYAYDC H.

PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Srsf7
Gene Accession No. Q16629, Q8BL97
Gene Alias 35kDa; 9430065L19Rik; 9G8; AAG3; aging-associated protein 3; hypothetical protein LOC507066; NX-96; serine and arginine rich splicing factor 7; serine/arginine-rich splicing factor 7; serine/arginine-rich splicing factor 7-like protein; SFRS7; Splicing factor 9G8; splicing factor, arginine/serine-rich 7; splicing factor, arginine/serine-rich 7, 35kDa; SR splicing factor 7; SRSF7
Gene Symbols Srsf7
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225027, 362687, 6432
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.