Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101911 Supplier Invitrogen™ Supplier No. PA5101911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SFRS2B.

The SR (serine/arginine-rich) family contains a number of phosphoproteins that function as essential and alternative splicing factors. The SR family of proteins is characterized by the presence of a ribonucleoprotein (RNP)-type RNA binding motif and a carboxyl-terminal arginine-serine-rich (RS) domain. SFRS2B is a member of the SR family and functions as an essential splicing factor in vitro.The SR (serine/arginine-rich) family contains a number of phosphoproteins that function as essential and alternative splicing factors. The SR family of proteins is characterized by the presence of a ribonucleoprotein (RNP)-type RNA binding motif and a carboxyl-terminal arginine-serine-rich (RS) domain. The protein encoded by this gene is a member of the SR family and functions as an essential splicing factor in vitro. This gene is thought to be an expressed PR264/SC35 retropseudogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SRSF8
Gene Accession No. Q9BRL6
Gene Alias DSM-1; pre-mRNA-splicing factor SRP46; serine and arginine rich splicing factor 8; serine/arginine-rich splicing factor 8; SFRS2B; Splicing factor SRp46; Splicing factor, arginine/serine-rich 2B; splicing factor, arginine/serine-rich, 46kD; SR splicing factor 8; SRP46; SRSF8
Gene Symbols SRSF8
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRSF8(Accession Q9BRL6), corresponding to amino acid residues R39-V89.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10929
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.