Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541776
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541776 Supplier Invitrogen™ Supplier No. PA541776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VCYADVQKDG VGMVEYLRKE DMEYALRKLD DTKFRSHEGE TSYIRVYPER Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

The protein encoded by this gene is a member of the serine/arginine (SR)-rich family of pre-mRNA splicing factors, which constitute part of the spliceosome. Each of these factors contains an RNA recognition motif (RRM) for binding RNA and an RS domain for binding other proteins. The RS domain is rich in serine and arginine residues and facilitates interaction between different SR splicing factors. In addition to being critical for mRNA splicing, the SR proteins have also been shown to be involved in mRNA export from the nucleus and in translation. Two pseudogenes, one on chromosome 15 and the other on chromosome 21, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SRSF9
Gene Accession No. Q13242
Gene Alias 25kDa; 2610029M16Rik; hypothetical protein zgc:77449; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 9; pre-mRNA-splicing factor SRp30C; serine and arginine rich splicing factor 9; serine/arginine-rich splicing factor 9; Sfrs9; splicing factor, arginine/serine rich 9; splicing factor, arginine/serine-rich 9; SR splicing factor 9; SRP30C; Srsf9; zgc:77449
Gene Symbols SRSF9
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SFRS9.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8683
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.