Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SRT 1720
GREENER_CHOICESDP

Catalog No. 755850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-585-0 50 mg
75-581-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-585-0 Supplier Tocris Bioscience™ Supplier No. 7558/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective SIRT1 activator; induces autophagy and mitochondrial biogenesis

SRT 1720 is selective SIRT 1 activator (EC50 = 0.16 μM). SRT 1720 inhibits growth of bladder cancer organoids and xenografts (IC50 = 0.35 μM) and induces autophagy. In vitro, SRT 1720 increases mitochondrial biogenesis and recovery after oxidation injury and reduces palmitate-induced insulin resistance. Also improves survival and healthspan of obese mice.

Specifications

Chemical Name or Material N-[2-[3-(1-Piperazinylmethyl)imidazo[2,1-b]thiazol-6-yl]phenyl]-2-quinoxalinecarboxamide
CAS 925434-55-5
Quantity 50 mg
Target Sirtuin 1/SIRT1
Molecular Formula C25H23N7OS
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.