Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRXN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106276 Supplier Invitrogen™ Supplier No. PA5106276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SRX1.

Contributes to oxidative stress resistance by reducing cysteine-sulfinic acid formed under exposure to oxidants in the peroxiredoxins PRDX1, PRDX2, PRDX3 and PRDX4. Does not act on PRDX5 or PRDX6. May catalyze the reduction in a multi-step process by acting both as a specific phosphotransferase and a thioltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRXN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SRXN1
Gene Accession No. Q9BYN0, Q9D975
Gene Alias 1700127B04Rik; Ab2-390; AI854065; AW488194; C20orf139; dJ850E9.2; neoplastic progression 3; neoplastic progression protein 3; Npn3; Srx; Srx1; SRXN1; sulfiredoxin 1; sulfiredoxin 1 homolog (S. cerevisiae); sulfiredoxin-1; TX01; YKL086W
Gene Symbols SRXN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRXN1(Accession Q9BYN0), corresponding to amino acid residues V41-D91.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140809, 296271, 76650
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.