Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRXN1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519037
Description
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Contributes to oxidative stress resistance by reducing cysteine-sulfinic acid formed under exposure to oxidants in the peroxiredoxins PRDX1, PRDX2, PRDX3 and PRDX4. Does not act on PRDX5 or PRDX6. May catalyze the reduction in a multi-step process by acting both as a specific phosphotransferase and a thioltransferase.
Specifications
| SRXN1 | |
| Polyclonal | |
| Unconjugated | |
| SRXN1 | |
| 1700127B04Rik; Ab2-390; AI854065; AW488194; C20orf139; dJ850E9.2; neoplastic progression 3; neoplastic progression protein 3; Npn3; Srx; Srx1; SRXN1; sulfiredoxin 1; sulfiredoxin 1 homolog (S. cerevisiae); sulfiredoxin-1; TX01; YKL086W | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 140809 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BYN0 | |
| SRXN1 | |
| Synthetic peptide sequence (KVQSLVDTIREDPD) corresponding to the internal amino acids of SRXN1 (aa 61-75). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction