Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRY Antibody (OTI3C8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274352H
Description
SRY Monoclonal specifically detects SRY in Human samples. It is validated for Western Blot.Specifications
SRY | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SRY | |
Full length recombinant protein of human SRY (NP_003131) produced in E.coli. | |
0.1 mL | |
6736.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3C8 | |
Western Blot | |
essential protein for sex determination in human males, sex determining region Y, sex-determining region on Y, sex-determining region Y protein, TDFsex determining region protein, TDY, Testis-determining factor | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction