Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRY Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5121576
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121576 100 μL
PIPA5121577 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121576 Supplier Invitrogen™ Supplier No. PA5121576
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This intronless gene encodes a transcription factor that is a member of the high mobility group (HMG)-box family of DNA-binding proteins. This protein is the testis-determining factor (TDF), which initiates male sex determination. Mutations in this gene give rise to XY females with gonadal dysgenesis (Swyer syndrome); translocation of part of the Y chromosome containing this gene to the X chromosome causes XX male syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRY
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SRY
Gene Accession No. Q05066
Gene Alias essential protein for sex determination in human males; HMG box transcription factor Sry1; HMG box transcription factor Sry3BI; sex determining region of Chr Y; sex determining region on Y; sex determining region Y; sex determining region Y protein; sex-determining region on Y; sex-determining region Y protein; SRXX1; SRXY1; Sry; Sry1; Sry3BI; SRYGENE; TDF; TDY; Testis determining locus; testis determining-Y; testis-determining factor; testis-determining factor on Y
Gene Symbols SRY
Host Species Rabbit
Immunogen Met1-Leu204 (Accession Q05066), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6736
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.