Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SRY Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101147
Description
Antibody detects endogenous levels of total SRY.
This intronless gene encodes a transcription factor that is a member of the high mobility group (HMG)-box family of DNA-binding proteins. This protein is the testis-determining factor (TDF), which initiates male sex determination. Mutations in this gene give rise to XY females with gonadal dysgenesis (Swyer syndrome); translocation of part of the Y chromosome containing this gene to the X chromosome causes XX male syndrome.
Specifications
| SRY | |
| Polyclonal | |
| Unconjugated | |
| SRY | |
| essential protein for sex determination in human males; HMG box transcription factor Sry1; HMG box transcription factor Sry3BI; sex determining region of Chr Y; sex determining region on Y; sex determining region Y; sex determining region Y protein; sex-determining region on Y; sex-determining region Y protein; SRXX1; SRXY1; Sry; Sry1; Sry3BI; SRYGENE; TDF; TDY; Testis determining locus; testis determining-Y; testis-determining factor; testis-determining factor on Y | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25221, 6736 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36394, Q05066 | |
| SRY | |
| A synthesized peptide derived from human SRY(Accession Q05066), corresponding to amino acid residues V60-F110. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction