Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperScript™ III CellsDirect™ cDNA Synthesis Kit

The SuperScript III CellsDirect cDNA Synthesis Kit is optimized for synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating the RNA.

Supplier:  Invitrogen™ 18080200

Encompass_Preferred

 View more versions of this product

Catalog No. 18-080-200


Only null left
Add to Cart

Description

Description

The SuperScript III CellsDirect cDNA Synthesis Kit is optimized for synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating the RNA. Lysis and reverse transcription are performed in the same tube, and the resulting first-strand cDNA is ready to use in cloning and PCR.

Features of the SuperScript III CellsDirect Synthesis Kit include:
• Compatible with a wide range of mammalian cell types grown under different treatment conditions
• Single-tube format minimizes reagent loss, sample loss, and handling time
• Total lysate volume is used in first-strand cDNA synthesis reaction, providing greater yields with a limited number of cells and allowing for detection of rare transcripts
• SuperScript III Reverse Transcriptase, with reduced RNase H activity and higher thermal stability, produces high yields of cDNA in the first-strand synthesis reaction, for greater sensitivity and enhanced detection of rare transcripts
• Generates high-quality cDNA for use in a variety of applications, including cloning and PCR
• Simple protocol takes less than two hours

How it works
In traditional RT-PCR, RNA is first isolated from cells in a time-consuming procedure that can lead to a loss of material. Using the SuperScript III CellsDirect cDNA Synthesis Kit, the cells are lysed and the cDNA is generated from the lysate in a single tube with minimal handling and no sample loss. DNase I is added to eliminate genomic DNA prior to first-strand synthesis. This kit has been optimized for small cell samples, ranging from 10,000 cells down to a single cell (as measured by serial dilution). The use of SuperScript III Reverse Transcriptase ensures high specificity and high yields of cDNA from small amounts of starting material (as little as 10 pg total RNA). After synthesis, the first-strand cDNA can be amplified with specific primers by PCR without intermediate organic extractions or ethanol precipitations.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• SuperScript III RT
• RNaseOUT Ribonuclease Inhibitor
• Oligo(dT)20 primers
• DNase I
• DNase I buffer
• EDTA
• dNTP mix
• 5X First-strand buffer
• DTT (0.1 M)
• RNase H
• Control RNA & primers
• Resuspension buffer

Store at –20°C.
Guaranteed stable for 6 months when properly stored.

High
Reverse Transcription
25 rxns
Reduced
First-Strand cDNA
Separate components
Up to 4.5 kb
Kit
50 min.
50°C
SuperScript III
Real Time PCR (qPCR)
25 Reactions
Reverse Transcription
Cells
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.