Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SS18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549678 Supplier Invitrogen™ Supplier No. PA549678
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous level of total SYT protein.

The transcriptional coactivator SYT (synovial translocation protein) contains a conserved amino terminal SNH domain and a carboxy terminal QPGY domain, which is a functioning transcriptional activating sequence. Synovial sarcoma translocation (SSX) proteins, including SSX1-5, are transcriptional repressors that contain a repressor domain in their carboxy termini. SSX proteins are localized to the nucleus and expressed in testis and several types of cancers and, therefore, they are classified as C/T (cancer/testis) antigens. The t(x;18) translocation results in the fusion of the amino terminus of SYT to the carboxy terminus of either SSX1 or SSX2; both fusions result in the production of transcriptional activators.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SS18
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SS18
Gene Accession No. Q15532, Q62280
Gene Alias D130059H17; MGC116875; Protein SSXT; Protein SYT; SS18; SS18, nBAF chromatin remodeling complex subunit; Ssxt; synovial sarcoma associated SS18-beta; synovial sarcoma associated SS18-delta; synovial sarcoma associated SS18-gamma; synovial sarcoma translocated to X chromosome protein; synovial sarcoma translocation; synovial sarcoma translocation, chromosome 18; synovial sarcoma, translocated to X chromosome; synovial sarcoma-associated Ss18-alpha; SYT
Gene Symbols SS18
Host Species Rabbit
Immunogen Peptide sequence around aa.14~18(G-E-I-T-P) derived from Human SYT.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268996, 361295, 6760
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.