Learn More
SSA1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302519A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the tripartite motif family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The encoded protein is part of the RoSSA ribonucleoprotein, which includes a single polypeptide and one of four small RNA molecules. The RoSSA particle localizes to both the cytoplasm and the nucleus. RoSSA interacts with autoantigens in patients with Sjogren syndrome and systemic lupus erythematosus. Alternatively spliced transcript variants for this gene have been described but the full-length nature of only one has been determined.Specifications
SSA1 | |
Polyclonal | |
Unconjugated | |
TRIM21 | |
TRIM21; tripartite motif-containing 21; RNF81, RO52, SSA, SSA1; 52kD Ro/SSA autoantigen; 52kDa ribonucleoprotein autoantigen SS-A/Ro; Sicca syndrome antigen A; Sjogren syndrome antigen A1 (52kD, ribonucleoprotein autoantigen SS-A/Ro); Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); tripartite motif protein TRIM21; tripartite motif protein 21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6737 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P19474 | |
TRIM21 | |
Between 425 and 475 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.