Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSB Antibody (OTI2C8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274356
Description
SSB Monoclonal specifically detects SSB in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
SSB | |
Monoclonal | |
LYOPH | |
Western Blot 1:1000-2000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 2 μg/500 μl | |
La autoantigen, La ribonucleoprotein, lupus La protein, member 3, Sjoegren syndrome type B antigen, Sjogren syndrome antigen B (autoantigen La), SS-B, SS-B/La protein | |
Mouse | |
46.8 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
OTI2C8 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
SSB | |
Full-length protein expressed in 293T cell transfected with human SSB expression vector | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
6741 | |
Human, Canine, Primate, Monkey | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction